Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lisofylline (racemate) | 6493-06-7 | 98.6% | 280.33 g/mol | C13H20N4O3 | 5MG
SDP

Catalog No. 5000238242
Encompass_Preferred

(±)-Lisofylline is a racemic anti-inflammatory small molecule for preclinical research. It inhibits generation of phosphatidic acid and free fatty acids and reduces release of pro-inflammatory cytokines in models of oxidative tissue injury and experimental sepsis; it is used in research including Type 1 diabetes. The material is a white to off-white solid supplied in small vial sizes for laboratory use.

  • Purity 98.6%.
  • Molecular weight 280.33 g/mol.
  • Chemical formula C13H20N4O3.
  • Soluble in DMSO (100 mg/mL); ultrasonic recommended.
  • Provided as a solid suitable for in vitro and preclinical studies.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-38242 Supplier Medchemexpress LLC Supplier No. HY1260425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Lisofylline is a racemic anti-inflammatory small molecule for preclinical research. It inhibits generation of phosphatidic acid and free fatty acids and reduces release of pro-inflammatory cytokines in models of oxidative tissue injury and experimental sepsis; it is used in research including Type 1 diabetes. The material is a white to off-white solid supplied in small vial sizes for laboratory use.

  • Purity 98.6%.
  • Molecular weight 280.33 g/mol.
  • Chemical formula C13H20N4O3.
  • Soluble in DMSO (100 mg/mL); ultrasonic recommended.
  • Provided as a solid suitable for in vitro and preclinical studies.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.