Learn More
Medchemexpress LLC Lisofylline (racemic) | 6493-06-7 | MFCD00871850 | >98.0% | 280.32 g/mol | C13H20N4O3 | 10MG
SDP
Catalog No. 5000235029
Encompass_Preferred
Lisofylline (racemic) is a small-molecule research reagent with anti-inflammatory and immunomodulatory activity. It is used in biochemical and cellular studies to probe inflammatory signaling and lipid metabolism. Supplied as a 10 mg analytical standard for laboratory research use only.
  • Racemic form of lisofylline.
  • Anti-inflammatory and immunomodulatory activity.
  • Used in studies of inflammatory signaling and lipid metabolism.
  • High purity >98.0%.
  • Molecular formula C13H20N4O3; molecular weight 280.32 g/mol.
  • Intended for research use only; not for human use.
Catalog No. 50-002-35029 Supplier Medchemexpress LLC Supplier No. HY12604210MG
Lisofylline (racemic) is a small-molecule research reagent with anti-inflammatory and immunomodulatory activity. It is used in biochemical and cellular studies to probe inflammatory signaling and lipid metabolism. Supplied as a 10 mg analytical standard for laboratory research use only.
  • Racemic form of lisofylline.
  • Anti-inflammatory and immunomodulatory activity.
  • Used in studies of inflammatory signaling and lipid metabolism.
  • High purity >98.0%.
  • Molecular formula C13H20N4O3; molecular weight 280.32 g/mol.
  • Intended for research use only; not for human use.