Learn More
Biotang Inc LISORB 250 ML, BP-256, 100 mM Lithium acetate, 50 mM Tris-HCl (pH 8.0), 1 mM EDTA, and 1 mM Sorbitol. Prepared in 18.2 megohms water and filtered through 1 micron filter.
Supplier:  Biotang Inc BTBB655
Encompass_Preferred
LISORB 250 ML, BP-256, 100 mM Lithium acetate, 50 mM Tris-HCl (pH 8.0), 1 mM EDTA, and 1 mM Sorbitol. Prepared in 18.2 megohms water and filtered through 1 micron filter.. This item has a minimum order quantity of 5 per supplier requirements for all the buffers.
Catalog No. 50-155-935