Learn More
NEOGEN Listeria Enrichment Broth, 500 g
SDP
Supplier:  NEOGEN 700003117
Encompass_Preferred
Listeria Enrichment Broth is used for selective enrichment of Listeria spp and is not intended for use in the diagnosis of disease or other conditions in humans.Listeria monocytogenes, first described in 1926 by Murray, Webb, and Swann, is an extensive problem in public health and food industries. This organism has the ability to cause human illness and death, particularly in immunocompromised individuals and pregnant women. Epidemiological evidence from outbreaks of listeriosis has indicated that the principle route of transmission is via consumption of foodstuffs contaminated with Listeria monocytogenes. Implicated vehicles of transmission include turkey frankfurters, coleslaw, pasteurized milk, Mexican style cheese, and pate. Listeria spp. are ubiquitous in nature, being present in a wide range of unprocessed foods as well as in soil, sewage, and river water.For more information visit https://www.neogen.com/solutions/Microbiology/listeria-enrichment-broth
Catalog No. 50-201-5312