Learn More
Seracare Life Sciences Inc LISTERIA GENUS POS CONTROL
Supplier:  Seracare Life Sciences Inc 53700010
Encompass_Preferred
Heat-killed Listeria monocytogenes cells. It consists of 1 mL of approximately 2% w/v wet packed cells in dextran solution. It is intended for use as a positive control in immunoassays designed for Listeria detection.
Catalog No. 50-674-62