Learn More
ATCC Listeria ivanovii; subsp. ivanovii
Supplier:  ATCC 19119
Encompass
Listeria ivanovii subsp. ivanovii strain Li 1979 is a whole-genome sequenced bacterial type strain isolated from sheep in Bulgaria. It has applications as a reference material and can be used as an indicator strain in the CAMP test.
Catalog No. 50-238-1706