Learn More
ATCC Listeria monocytogenes
Supplier:  ATCC 13932
Encompass
Listeria monocytogenes strain 1071/53 [LMG 21264, NCTC 10527] was isolated from the spinal fluid of a child with meningitis. This bacterium has applications in enteric disease research and media testing.
Catalog No. 50-238-2746