Learn More
Sino Biological Recombinant Anti-flagellin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 40067R121100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Listeria monocytogenes Listeria monocytogenes flagellin / flaA (Catalog#40067-V07E; YP_013330.1; Met1-Ser287).
Catalog No. 50-237-3645