Learn More
ATCC Listeria monocytogenes; genomic DNA
Supplier:  ATCC 19115D5
Encompass
Genomic DNA isolated from Listeria monocytogenes strain Li 2 (ATCC 19115). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3444