Learn More
ATCC Listeria monocytogenes; Strain: LMP18-H8393
Supplier:  ATCC BAA3153
Encompass
Listeria monocytogenes strain LMP18-H8393 was isolated from chocolate milk. This strain was part of the 2018 PulseNet-GenomeTrakr joint proficiency test exercise. It is typed as serotype 1/2b and has applications in food testing and enteric disease research.
Catalog No. 50-238-4572