Learn More
AdipoGen anti-Listeria monocytogenes p60, mAb (P6007) (100 µg)
SDP
Supplier:  AdipoGen AG20A0022C100
Encompass_Preferred
Monoclonal Antibody. Recognizes L. monocytogenes p60 protein. Does not cross-react with other Listeria species. Isotype: Mouse IgG1kappa. Clone: p6007. Applications: ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Listeria monocytogenes is a intracellular bacterium that is the causative agent of Listeriosis. It is one of the most virulent foodborne pathogens. Listeria species are known to secrete a 60-kDa protein collectively termed p60, which is encoded by the iap (invasion-associated-protein) gene and secreted in large quantites into the growth media. p60 is a highly immunogenic murein hydrolase that is essential for cell division.
Catalog No. 50-205-2801