Learn More
Stoelting™ Liston Bone Cutter
SDP
Catalog No. 10001810 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
5.5 in.
8.5 in.
Model:
Angled
Staight
Straight
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Model
10-001-810 8.5 in. Angled
10-001-807 5.5 in. Straight
10-001-809 5.5 in. Angled
10-001-808 8.5 in. Staight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-001-810 Supplier Stoelting™ Supplier No. 5216779
Add to Cart
Add to Cart
Bone cutting tools allow for sharp cuts through soft bone and cartilage.
Specifications
For Use With (Application) Neuroscience
Length (English) 8.5 in.
Model Angled