Learn More
Electron Microscopy Sciences Liston Bone Cutting Forceps with Straight Blades, 8.5"
SDP
Catalog No. 5030195
Encompass_Preferred
Box joint bone cutting forceps. Made from stainless steel with a polished finish.
OAL: 8½"
Catalog No. 50-301-95 Supplier Electron Microscopy Sciences Supplier No. 779948S
Box joint bone cutting forceps. Made from stainless steel with a polished finish.
OAL: 8½"