Learn More
Sklar Instruments Stille-Liston Bone Cutting Forceps, Sklar, 9 in., Straight, Stainless Steel, Double Action. EA/1.
SDP
Catalog No. 501438554 Shop All Sklar Instruments Products
Encompass_Preferred
Stille-Liston Bone Cutting Forceps, Sklar, 9 in., Straight, Stainless Steel, Double Action. Sklar's Stille-Liston Bone Cutting Forceps are used for cutting bones. It is a double-action instrument that provides greater pressure on the bones without as much effort by the user. The jaws of the forceps have sharp blades on the inside edge and are rounded on the outer edge. This product is straight and has an overall length of 9 inches.
This item is not returnable. View return policy
Catalog No. 50-143-8554 Supplier Sklar Instruments Supplier No. 403709
Stille-Liston Bone Cutting Forceps, Sklar, 9 in., Straight, Stainless Steel, Double Action. Sklar's Stille-Liston Bone Cutting Forceps are used for cutting bones. It is a double-action instrument that provides greater pressure on the bones without as much effort by the user. The jaws of the forceps have sharp blades on the inside edge and are rounded on the outer edge. This product is straight and has an overall length of 9 inches.