Learn More
Mopec™ Liston Bone Cutting Straight, Standard Forceps
SDP
Catalog No. 22445431 Shop All Mopec Products
Change view
Click to view available options
Length (English):
8.5 in.
Catalog No. Length (English)
22-445-431 8.5 in.
1 options
Catalog No. 22-445-431 Supplier Mopec™ Supplier No. AG051
Add to Cart
Add to Cart
Effective bone cutting instrument for use during autopsies and post-mortem dissection. Sharp tips perform a clean extraction.
  • Complete with smooth grip
Specifications
Autoclavable Not Autoclavable
Color Silver
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Length (English) 8.5 in.