Learn More
Med Vet International Miltex Vantage Liston Bone Cutting Forceps, 5.5in., Straight
Catalog No. 502830185
Encompass_Preferred
The Miltex Vantage V927-1390 Liston bone cutting forceps are made from high quality Japanese stainless steel and measure 5.5" in length. They have a non-locking plier handle with a spring, and straight jaws.
Catalog No. 50-283-0185 Supplier Med Vet International Supplier No. MILV9271390
The Miltex Vantage V927-1390 Liston bone cutting forceps are made from high quality Japanese stainless steel and measure 5.5" in length. They have a non-locking plier handle with a spring, and straight jaws.