Learn More
Surgical Design™ Liston-Stille Bone Cutting Rongeur
Catalog No. 21124473 Shop All Surgical Design Products
Change view
Click to view available options
Blade Style:
Straight
Catalog No. Blade Style
21-124-473 Straight
1 options
Catalog No. 21-124-473 Supplier Surgical Design™ Supplier No. 111335
Add to Cart
Add to Cart
Liston-Stille bone cutting rongeur delivers exceptional strength and cutting precision with a 10.25 in. stainless-steel design for bone sectioning.
  • Constructed from medical-grade stainless steel for maximum strength, durability, and corrosion resistance
  • 10.25-in. length provides superior reach and leverage for large bone cutting tasks
  • Double-action mechanism enhances cutting power with smooth, controlled operation
  • Heavy-duty jaws deliver clean, efficient sectioning of dense or cortical bone
  • Precision-ground cutting edges ensure consistent and reliable performance
  • Ergonomic handle design provides a secure grip and reduces hand fatigue during prolonged procedures
  • Balanced weight distribution allows for precise handling and control
  • Commonly used in autopsy, forensic, and pathology bone dissection
  • Suitable for veterinary and research settings requiring high-strength cutting instruments
Specifications
Product Type Bone-cutting Forceps
Blade Material Stainless Steel
Blade Style Straight
Length (English) 10.25 in.
Length (Metric) 26 cm
Material Stainless Steel
Autoclavable Autoclavable