Learn More
Medchemexpress LLC Lisuride maleate | 19875-60-6 | 454.52 | 1 ML
SDP
Catalog No. 5000345441
Encompass_Preferred
Lisuride maleate is a potent dopamine agonist that directly acts on dopaminergic receptors. This ergot derivative can release premenstrual mastalgia without significant side effects. Research indicates its in vivo potency, demonstrating effectiveness in antagonizing motor depression and hypothermia. It has also been involved in clinical trials for conditions such as Parkinson's Disease, Restless Legs Syndrome, and Cocaine-Related Disorders.
  • Potent dopamine agonist
  • Releases premenstrual mastalgia
  • Antagonizes motor depression and hypothermia
  • Used in clinical trials for neurological disorders
  • Stable under recommended storage conditions
  • Soluble in various solvents for research applications
Catalog No. 50-003-45441 Supplier Medchemexpress LLC Supplier No. HY11008010MG
Add to Cart
Add to Cart
Lisuride maleate is a potent dopamine agonist that directly acts on dopaminergic receptors. This ergot derivative can release premenstrual mastalgia without significant side effects. Research indicates its in vivo potency, demonstrating effectiveness in antagonizing motor depression and hypothermia. It has also been involved in clinical trials for conditions such as Parkinson's Disease, Restless Legs Syndrome, and Cocaine-Related Disorders.
  • Potent dopamine agonist
  • Releases premenstrual mastalgia
  • Antagonizes motor depression and hypothermia
  • Used in clinical trials for neurological disorders
  • Stable under recommended storage conditions
  • Soluble in various solvents for research applications