Learn More
Medchemexpress LLC Lisuride maleate | 19875-60-6 | 99.8% | 454.52 | 1 MG
SDP
Catalog No. 5000345099
Encompass_Preferred
Lisuride is a potent dopamine agonist, acting directly on dopaminergic receptors. It is an ergot derivative that has been shown to alleviate premenstrual mastalgia. It is intended for research use only.
  • Potent agonist of dopamine with a direct action on dopaminergic receptors
  • Ergot derivative
  • Releases premenstrual mastalgia without significant side effects
Catalog No. 50-003-45099 Supplier Medchemexpress LLC Supplier No. HY11008010MM/1ML
Lisuride is a potent dopamine agonist, acting directly on dopaminergic receptors. It is an ergot derivative that has been shown to alleviate premenstrual mastalgia. It is intended for research use only.
  • Potent agonist of dopamine with a direct action on dopaminergic receptors
  • Ergot derivative
  • Releases premenstrual mastalgia without significant side effects