Learn More
Medchemexpress LLC Lisuride maleate | 19875-60-6 | 99.97% | 454.52 | 50 MG
SDP
Catalog No. 5000343470
Encompass_Preferred
Lisuride (maleate) is a potent dopamine agonist that is believed to act directly on dopaminergic receptors. It is an ergot derivative. This compound has been shown to alleviate premenstrual mastalgia without significant side effects. Lisuride (maleate) has been studied in vivo, demonstrating 3-4 times higher potency when administered intraperitoneally (i.p.) compared to orally (p.o.). It effectively antagonizes motor depression and hypothermia induced by reserpine. At dosages as low as 0.10 mg/kg i.p., it is comparable in effectiveness to apomorphine and D-amphetamine in causing significant hypothermia. The product is for research use only and not sold to patients.
  • Potent dopamine agonist
  • Ergot derivative
  • Therapeutic potential for premenstrual mastalgia
  • Suitable for research applications
  • High purity
  • Versatile formulations
Catalog No. 50-003-43470 Supplier Medchemexpress LLC Supplier No. HY1100801MG
Lisuride (maleate) is a potent dopamine agonist that is believed to act directly on dopaminergic receptors. It is an ergot derivative. This compound has been shown to alleviate premenstrual mastalgia without significant side effects. Lisuride (maleate) has been studied in vivo, demonstrating 3-4 times higher potency when administered intraperitoneally (i.p.) compared to orally (p.o.). It effectively antagonizes motor depression and hypothermia induced by reserpine. At dosages as low as 0.10 mg/kg i.p., it is comparable in effectiveness to apomorphine and D-amphetamine in causing significant hypothermia. The product is for research use only and not sold to patients.
  • Potent dopamine agonist
  • Ergot derivative
  • Therapeutic potential for premenstrual mastalgia
  • Suitable for research applications
  • High purity
  • Versatile formulations