Learn More
Medchemexpress LLC Lisuride (maleate) | 19875-60-6 | MFCD28166478 | 100.0% | 454.52 g·mol⁻¹ | C24H30N4O5 | 5 MG
SDP
Catalog No. 5000264835
Encompass_Preferred
Lisuride (maleate) is an ergot-derivative dopamine D2 receptor agonist supplied as a white to off-white solid for research use. It is commonly used in preclinical pharmacology studies of dopaminergic signaling, Parkinson's disease models, and migraine research, and is provided with characterized physicochemical properties for compound identification.
  • High purity suitable for analytical and preclinical research.
  • White to off-white solid with defined storage recommendations to maintain stability.
  • Soluble in DMSO at approximately 25 mg/mL with warming and sonication.
  • Characterized molecular weight and formula for reliable identification.
  • Available in small pack sizes for screening and dosing studies.
Catalog No. 50-002-64835 Supplier Medchemexpress LLC Supplier No. HY1100805MG
Lisuride (maleate) is an ergot-derivative dopamine D2 receptor agonist supplied as a white to off-white solid for research use. It is commonly used in preclinical pharmacology studies of dopaminergic signaling, Parkinson's disease models, and migraine research, and is provided with characterized physicochemical properties for compound identification.
  • High purity suitable for analytical and preclinical research.
  • White to off-white solid with defined storage recommendations to maintain stability.
  • Soluble in DMSO at approximately 25 mg/mL with warming and sonication.
  • Characterized molecular weight and formula for reliable identification.
  • Available in small pack sizes for screening and dosing studies.