Learn More
Cayman Chemical Lisuride (maleate), Purity: ≥98%, CAS: 19875-60-6, 5 mg
SDP
Catalog No. 502911718
Encompass_Preferred
An analytical reference standard categorized as a lysergamide; does not induce the HTR in mice, indicating a lack of hallucinogenic potential; intended for research and forensic applications
Catalog No. 50-291-1718 Supplier Cayman Chemical Supplier No. 361405 MG
An analytical reference standard categorized as a lysergamide; does not induce the HTR in mice, indicating a lack of hallucinogenic potential; intended for research and forensic applications