Learn More
Selleck Chemical LLC LIT-001 trifluoroacetate
SDP
Supplier:  Selleck Chemical LLC E06205MG
Encompass_Preferred
LIT-001 trifluoroacetate(LIT-001 TFA) is the first nonpeptide oxytocin receptor (OXTR) agonist that improves social interaction in a mouse model of autism
Catalog No. 50-313-7618