Learn More
Honeywell™ Lite Force Plus Structural Helmets
Catalog No. 19167100
Change view
Click to view available options
Color:
Black
Red
White
Yellow
Includes:
Yellow Nomex/Crosstech PJ/FR cotton ear covers
Yellow Nomex/FR cotton ear covers
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
19-167-100 Yellow Yellow Nomex/FR cotton ear covers
19-167-149 White Yellow Nomex/Crosstech PJ/FR cotton ear covers
19-167-098 Red Yellow Nomex/FR cotton ear covers
19-167-148 Red Yellow Nomex/Crosstech PJ/FR cotton ear covers
19-167-147 Black Yellow Nomex/Crosstech PJ/FR cotton ear covers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-167-100 Supplier Honeywell™ Supplier No. SAEGSA00525
Add to Cart
Add to Cart
Rugged and dependable with FYR-glass technology
  • No assembly required
  • Clear 4 × 0.110 in. APEC high-temperature faceshield
  • Nomex™ quick-release chinstrap with postman slide
  • Five 1 × 4 in. Reflexite™ lime bars
  • Kevlar™ felt winter liner dome
  • Choice of yellow Nomex/FR cotton or Nomex/Crosstech™ PJ/FR cotton ear covers
  • Order Info
    Available through Fisher Safety's GSA Advantage!™ Schedule 84.
    Compliance
    Meet NFPA 1971, 2007 Edition.
    Warranty and Services
    Lifetime warranty
    TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance Meets NFPA 1971, 2007 Edition.
    Color Yellow
    Includes Yellow Nomex/FR cotton ear covers