Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lith-O-Asp | 881179-02-8 | MFCD31692386 | 98.0% | 491.66 g/mol | C28H45NO6 | 5 MG
SDP

Catalog No. 5000264697
Encompass_Preferred

Lith-O-Asp is a small-molecule sialyltransferase (ST) inhibitor reported to inhibit ST activity with IC50 values of 12-37 μM. It is provided as a purified research reagent in solid form for biochemical and cell-based studies; not for human therapeutic use.

  • Inhibits sialyltransferase with IC50 12-37 μM.
  • High purity (98.0%).
  • Molecular formula C28H45NO6 and molecular weight 491.66 g/mol.
  • Supplied as a solid in milligram quantities suitable for screening and assays.
  • Intended for research use only; not for therapeutic applications.
  • CAS number 881179-02-8 for unambiguous identification.

Catalog No. 50-002-64697 Supplier Medchemexpress LLC Supplier No. HY1124155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lith-O-Asp is a small-molecule sialyltransferase (ST) inhibitor reported to inhibit ST activity with IC50 values of 12-37 μM. It is provided as a purified research reagent in solid form for biochemical and cell-based studies; not for human therapeutic use.

  • Inhibits sialyltransferase with IC50 12-37 μM.
  • High purity (98.0%).
  • Molecular formula C28H45NO6 and molecular weight 491.66 g/mol.
  • Supplied as a solid in milligram quantities suitable for screening and assays.
  • Intended for research use only; not for therapeutic applications.
  • CAS number 881179-02-8 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.