Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide 98.0+%, TCI America™
SDP

Catalog No. H10575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10575G 5 g
H10571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10575G Supplier TCI America Supplier No. H10575G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 189217-62-7
Molecular Formula C3F6LiNO4S2
Molecular Weight (g/mol) 299.086
MDL Number MFCD06200805
InChI Key VKQJSJGVEJXINS-UHFFFAOYSA-N
Synonym 4,4,5,5,6,6-Hexafluoroperhydro-1,3,2-dithiazine 1,1,3,3-Tetraoxide Lithium Salt, 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide Lithium Salt
PubChem CID 44630347
IUPAC Name lithium;4,4,5,5,6,6-hexafluoro-1$l^{6},3$l^{6}-dithia-2-azanidacyclohexane 1,1,3,3-tetraoxide
SMILES [Li+].C1(C(S(=O)(=O)[N-]S(=O)(=O)C1(F)F)(F)F)(F)F

Specifications

Color White
Quantity 5 g
Formula Weight 299.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Lithium 1,1,2,2,3,3-Hexafluoropropane-1,3-disulfonimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.