Learn More
SPEX CERTIPREP LLC 3P LITHIUM 1000UG/ML STD 100ML
Catalog No. NC0664536
Encompass_Preferred
(HPS by Zeptometrix) Li 1000ug/mL in 1% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0664536 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000291100
(HPS by Zeptometrix) Li 1000ug/mL in 1% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.