Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium 2,2,6,6-Tetramethylpiperidide

Catalog No. 73535325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-535-325G 25 g
73-535-310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 73-535-325G Supplier Sigma Aldrich Supplier No. 73535325G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium 2,2,6,6-tetramethylpiperidide is a strong base and can be used:For the synthesis of enamines from terminal epoxides through trans-+a-lithiated epoxide as an intermediate.For ortholithiation of arenes such as 1,3-bis(trifluoromethyl)benzene, 4,4-dimethyl-2-phenyl-2-oxazoline, 1,4-bis(trifluoromethyl)benzene and 1,3-dimethoxybenzene.In combination with Lewis donor ligand, N,N,N'sa,N'sa-tetramethylethylenediamine (TMEDA) for deprotonative metalation of methoxy-substituted arenes.{21]

Specifications

CAS 38227-87-1
Molecular Formula C9H18LiN
Linear Formula C9H18LiN
MDL Number MFCD00015910
Quantity 25 g
Synonym 2,2,6,6-Tetramethylpiperidine lithium salt (1:1); LTMP
Molecular Weight (g/mol) 147.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.