Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 2,4-pentanedionate, 99.5%

Catalog No. AA3052918
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3052918 50 g
AA3052909 10 g
AA3052930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3052918 Supplier Thermo Scientific Chemicals Supplier No. 03052918
Only null left
Add to Cart

CAS: 18115-70-3 | C5H7LiO2 | 106.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18115-70-3
Molecular Formula C5H7LiO2
Molecular Weight (g/mol) 106.05
MDL Number MFCD00013496
InChI Key JTEOOCAGEXVCBQ-LNKPDPKZSA-M
Synonym Lithium acetylacetonate
IUPAC Name lithium(1+) (2Z)-4-oxopent-2-en-2-olate
SMILES [Li+].C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼250°C (decomposition)
Color White
Quantity 50 g
Beilstein 4575087
Sensitivity Hygroscopic
Formula Weight 106.05
Percent Purity 99.5%
Physical Form Crystalline
Chemical Name or Material Lithium 2,4-pentanedionate
Hazard Category H302+H312+H332-H315-H319-H335-H361
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 242-008-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.