Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lithium 2-hydroxypropanoate | 867-55-0 | 99.9% | 96.01 | 50 G
SDP

Catalog No. 5000453606
Encompass_Preferred

Lithium 2-hydroxypropanoate, also known as Propanoic acid, 2-hydroxy-, lithium salt (1:1), is a solid chemical supplied by MedChemExpress for laboratory use and the manufacture of substances. It is stable under recommended storage conditions.

  • Storage at 4°C, sealed, away from moisture
  • White to off-white solid appearance
  • Purity (HPLC) 99.91%

Catalog No. 50-004-53606 Supplier Medchemexpress LLC Supplier No. HYW01062550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lithium 2-hydroxypropanoate, also known as Propanoic acid, 2-hydroxy-, lithium salt (1:1), is a solid chemical supplied by MedChemExpress for laboratory use and the manufacture of substances. It is stable under recommended storage conditions.

  • Storage at 4°C, sealed, away from moisture
  • White to off-white solid appearance
  • Purity (HPLC) 99.91%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.