Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 3,5-diiodosalicylate, MilliporeSigma™ Supelco™

Catalog No. 111015078 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5078 5 g
11-101-5077 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5078 Supplier MilliporeSigma Supelco Supplier No. D36355G
Add to Cart
Add to Cart

Specifications

Packaging Poly Bottle
CAS 653-14-5
Physical Form Neat
Linear Formula C7H3I2 LiO3
MDL Number MFCD00002443
Synonym 2-Hydroxy-3,5-diiodobenzoic acid lithium salt; 3,5-Diiodo-2-hydroxybenzoic acid lithium salt; 3,5-Diiodosalicylic acid lithium salt; LIS
Type Analytical Standard
Quantity 5 g
Formula Weight 395.85
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.