Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole, 95%

Catalog No. AAH3408103
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3408103 1 g
AAH3408109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3408103 Supplier Thermo Scientific Chemicals Supplier No. H3408103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 761441-54-7 | C6F3LiN4 | 192.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761441-54-7
Molecular Formula C6F3LiN4
Molecular Weight (g/mol) 192.03
MDL Number MFCD18827480
InChI Key CVVIFWCYVZRQIY-UHFFFAOYSA-N
Synonym litdi, lithium 4,5-dicyano-2-trifluoromethyl imidazole, lithium 4,5-dicyano-2-trifluoromethyl-imidazolide, 1-lithio-2-trifluoromethyl-1h-imidazole-4,5-dicarbonitrile
PubChem CID 73995168

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 192.03
Percent Purity 95%
Chemical Name or Material Lithium 4,5-dicyano-2-(trifluoromethyl)imidazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 691-963-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.