Learn More
Medchemexpress LLC Lithium acetate dihy 50g
SDP
Catalog No. 5000206541
Encompass_Preferred
Lithium acetate dihydrate is a compound with a one-dimensional structure that has various applications in industries such as pharmaceuticals ceramics and research laboratories Lithium acetate dihydrate is often used as a source of lithium ions in chemical reactions and as a precursor in the synthesis of other lithium compounds[1]
Catalog No. 50-002-06541 Supplier Medchemexpress LLC Supplier No. HYW09447150G
Lithium acetate dihydrate is a compound with a one-dimensional structure that has various applications in industries such as pharmaceuticals ceramics and research laboratories Lithium acetate dihydrate is often used as a source of lithium ions in chemical reactions and as a precursor in the synthesis of other lithium compounds[1]