Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium acetate dihydrate1KG
Catalog No. 502902345
Encompass
Lithium acetate dihydrate is one of the commercially available forms of lithium acetate. Lithium acetate is a common salt with variety of applications such as product for efficient yeast transformation drug formulation and therapy lithium-6 CP/MAS standard solvent and catalyst. In addition it is also used as a buffer for gel electrophoresis of DNA and RNA additive or catalyst for textiles and polymer production and as a precursor material for batteries and ferromagnetic nanoparticles.
Catalog No. 50-290-2345 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L41581KG
Lithium acetate dihydrate is one of the commercially available forms of lithium acetate. Lithium acetate is a common salt with variety of applications such as product for efficient yeast transformation drug formulation and therapy lithium-6 CP/MAS standard solvent and catalyst. In addition it is also used as a buffer for gel electrophoresis of DNA and RNA additive or catalyst for textiles and polymer production and as a precursor material for batteries and ferromagnetic nanoparticles.