Learn More
Selleck Chemical LLC Lithium acetoacetate S3353-25mg
SDP
Catalog No. 503108768
Encompass_Preferred
Lithium acetoacetate (LiAcAc Acetoacetic acid lithium salt Lithium 3-oxobutanoate) is an active endogenous metabolite that affects the growth of cells Lithium acetoacetate can be used as an acetoacetate standard for the measurement of acetoacetate in biological samples
Catalog No. 50-310-8768 Supplier Selleck Chemical LLC Supplier No. S335325MG
Lithium acetoacetate (LiAcAc Acetoacetic acid lithium salt Lithium 3-oxobutanoate) is an active endogenous metabolite that affects the growth of cells Lithium acetoacetate can be used as an acetoacetate standard for the measurement of acetoacetate in biological samples