Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium acetoacetate

Catalog No. A85095G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A85095G 5 g
A850910MG 10 mg
A85091G 1 g
A8509250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A85095G Supplier Sigma Aldrich Supplier No. A85095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Lithium acetoacetate can be used as an acetoacetate standard for the measurement of acetoacetate in biological samples
  • It can also be used for the preparation of sodium acetoacetate.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • A8509.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

Quantity 5 g
Molecular Formula C4H5LiO3
Linear Formula CH3COCH2COOLi
MDL Number MFCD00013060
Synonym Acetoacetic acid lithium salt
Molecular Weight (g/mol) 108.02
Percent Purity ≥90% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.