Learn More
Strem, An Ascensus Company CAS# 18115-70-3. 50g. Lithium acetylacetonate, 98+%. MFCD00013496
SDP
Catalog No. 5090113659
Encompass
CAS# 18115-70-3. 50g. Lithium acetylacetonate, 98+%. MFCD00013496. Molecular Weight: 106.05. Molecular Formula: LiCH3C(O)CHC(O)CH3. Color/form: white pwdr. Strem# 93-0361. http://www.strem.com/catalog/v/93-0361/
Catalog No. 50-901-13659 Supplier Strem, An Ascensus Company Supplier No. 93036150G
CAS# 18115-70-3. 50g. Lithium acetylacetonate, 98+%. MFCD00013496. Molecular Weight: 106.05. Molecular Formula: LiCH3C(O)CHC(O)CH3. Color/form: white pwdr. Strem# 93-0361. http://www.strem.com/catalog/v/93-0361/