Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium aluminium hydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
16853-85-3,109-99-9
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride,lithium aluminium hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
16853-85-3,109-99-9
AlH4Li
37.954
MFCD00011075
BJKLPLABXHXMIM-UHFFFAOYSA-N
lithium aluminum hydride,lithium aluminium hydride,aluminum lithium hydride,lithiumaluminiumhydride,aluminum iii lithium hydride,lithium alanate,lithium aluminum tetrahydride,lithiumaluminiumhydrid,litiumaluminum hydride,lithim aluminum hydride
21226445
aluminum;lithium;hydride
[H-].[H-].[H-].[H-].[Li+].[Al+3]
Reduction reagent
Linear Formula LiAlH4
Fieser 01,581; 02,242; 03,176; 04,291; 05,382; 06,325; 07,196; 08,286; 09,274; 10,236; 11,289; 12,272; 13,61; 14,190; 15,184; 16,133; 17,162
Merck Index 15, 344
Density 0.9000g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 37.95
Physical Form Turbid Solution
Percent Purity 3.9 to 4.5% (as LiAlH4)
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 0.9
Color Gray
Quantity 100mL
Flash Point −17°C
Chemical Name or Material Lithium aluminium hydride
Show More Show Less

GHS H Statement
May cause respiratory irritation. Causes skin irritation. Causes serious eye damage. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger