Learn More
Strem, An Ascensus Company CAS# 14128-54-2. 5g. Lithium aluminum deuteride, 98% isotopic purity. MFCD00044245
SDP
Supplier:  Strem, An Ascensus Company 0307505G
Encompass
CAS# 14128-54-2. 5g. Lithium aluminum deuteride, 98% isotopic purity. MFCD00044245. Molecular Weight: 41.99. Molecular Formula: LiAlD4. Color/form: gray pwdr. Strem# 03-0750. http://www.strem.com/catalog/v/03-0750/
Catalog No. 50-201-6668