Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Aluminum Hydride Solution

Catalog No. 236055100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-605-5100ML 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 23-605-5100ML Supplier Sigma Aldrich Supplier No. 236055100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant or reagent for: The preparation of thermoplastic polyester polyamides from oleic acidLithium-polymer batteriesHydrodefluorination of gem-difluoromethylene derivativesAsymmetric aldol reactionsSynthesis of Li-Al-N-H composites with hydrogen absorption / desorption propertiesLAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcohols

Specifications

CAS 7397-43-5
Melting Point 18°C to 19°C
Density 0.811 g/mL at 25°C
Boiling Point 101°C (74 mmHg, lit.)
Molecular Formula C12H27BO3
Linear Formula [(CH3)3CO]3B
MDL Number MFCD00009652
Quantity 50 g
Synonym LAH; Lithium alanate; Lithium tetrahydroaluminate
Molecular Weight (g/mol) 230.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.