Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Aluminum Hydride Solution

Catalog No. 59370225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
4 x 25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-370-225ML 4 x 25 mL
59-370-21L -
59-370-2100ML -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 59-370-225ML Supplier Sigma Aldrich Supplier No. 5937024X25ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant or reagent for: The preparation of thermoplastic polyester polyamides from oleic acidLithium-polymer batteriesHydrodefluorination of gem-difluoromethylene derivativesAsymmetric aldol reactionsSynthesis of Li-Al-N-H composites with hydrogen absorption / desorption propertiesLAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcoholsSure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 16853-85-3
Density 0.906 g/mL at 25°C
Quantity 4 x 25 mL
Molecular Formula H4AlLi
Linear Formula LiAlH4
MDL Number MFCD00011075
Synonym LAH; Lithium alanate; Lithium tetrahydroaluminate
Molecular Weight (g/mol) 37.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.