Learn More
Sigma Aldrich Lithium Aluminum Titanium Phosphate (latp) Powder Battery Grade
Catalog No. 91539450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
50 g
Catalog No. Quantity
91-539-450G 50 g
1 options
Catalog No. 91-539-450G Supplier Sigma Aldrich Supplier No. 91539450G
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Solid electrolytes (SEs) have become a practical option for lithium ion and lithium metal batteries because of their improved safety over commercially available ionic liquids. Solid electrolyte materials are highly conductive and made for advanced solid-state lithium ion battery technologies. These solutions are high purity and battery grade thus making them suitable as standards in LIB research.
Specifications
Quantity 50 g
Linear Formula Li1.4 Al0.4 Ti1.6(PO4)3