Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium amide, 95%

Catalog No. AAL1602336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1602336 500 g
AAL1602314 25 g
AAL1602322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1602336 Supplier Thermo Scientific Chemicals Supplier No. L1602336
  • $347.00
    $305.65 / Each of 1

    Save $41.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7782-89-0 | H2LiN | 22.963 g/mol

Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

Solubility
Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether, benzene and toluene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, water and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-89-0
Molecular Formula H2LiN
Molecular Weight (g/mol) 22.963
MDL Number MFCD00011093
InChI Key AFRJJFRNGGLMDW-UHFFFAOYSA-N
Synonym lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder
PubChem CID 24532
IUPAC Name lithium;azanide
SMILES [Li+].[NH2-]

Specifications

Boiling Point 430°C
Melting Point 373°C
Quantity 500 g
Linear Formula LiNH2
UN Number UN1390
Merck Index 14,5522
Solubility Information Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether,benzene and toluene.
Formula Weight 22.96
Percent Purity 95%
Sensitivity Air and moisture sensitive
Density 1.178
Chemical Name or Material Lithium amide
Show More Show Less
Hazard Category H261-H314
Hazard Statement GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ALKALI METAL AMIDES
EINECSNumber 231-968-4
RTECSNumber OJ5590000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.