Learn More
Crystal Chem Inc Lithium Assay Kit, 96-tests
SDP
Supplier:  Crystal Chem Inc 80159
Encompass
The Lithium Assay Kit is used to quantify levels of lithium in serum using only a 5 µL sample volume. Highlights: Convenient and liquid stable. Highly stable enzymatic method. High quality cost effective alternative to high priced dye binding.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7668