Learn More
Grainger 1/2 AA Battery, 3.6VDC, Lithium, Button, 1200mAh
Catalog No. NC1566618
Encompass_Preferred
1/2 AA batteries provide stable voltage for supplying memory backup in a device or for powering meter readers, alarms, tracking systems, and automotive electronics. Sometimes called 14250 batteries, they have a nominal diameter of 14.55 mm.
Catalog No. NC1566618 Supplier Grainger Supplier No. 20PD32
1/2 AA batteries provide stable voltage for supplying memory backup in a device or for powering meter readers, alarms, tracking systems, and automotive electronics. Sometimes called 14250 batteries, they have a nominal diameter of 14.55 mm.