Learn More
Lithium bis(trimethylsilyl)amide, 1M solution in THF/Ethylbenzene, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 347701000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium bis(trimethylsilyl)amide | |
4039-32-1, 109-99-9, 100-41-4 | |
0.8900g/mL | |
AcroSeal™ Glass bottle | |
100 mL | |
04,296; 05,393; 07,197; 12,280; 13,165; 14,194 | |
lithium bis trimethylsilyl amide, lithium hexamethyldisilazide, lihmds, lhmds, hexamethyldisilazane lithium salt, unii-rc4n1i108m, lithiumbis trimethylsilyl amide, lithium bis trimethylsilyl azanide, lithium hexamethyldisilazane, lithium bis-trimethylsilyl amide | |
YNESATAKKCNGOF-UHFFFAOYSA-N | |
lithium;bis(trimethylsilyl)azanide | |
2733832 | |
18 to 22% active base (as LiNSi) |
1M Solution in THF/Ethylbenzene | |
Yellow to Brown | |
−21°C | |
C6H18LiNSi2 | |
MFCD00008261 | |
0.89 | |
Solubility in water: decomposes | |
[Li+].C[Si](C)(C)[N-][Si](C)(C)C | |
167.33 | |
167.33 | |
Turbid Solution |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Suspected of causing cancer.
Reacts violently with water.
May form explosive peroxides.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha
GHS Signal Word: Danger
EINECSNumber : 223-725-6
TSCA : TSCA
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only