Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Bis(Trimethylsilyl)Amide

Catalog No. 324620250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-462-0250G 250 g
32-462-050G 50 g
32-462-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 32-462-0250G Supplier Sigma Aldrich Supplier No. 324620250G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium bis(trimethylsilyl)amide is a non-nucleophilic strong Br++nsted base, which is generally soluble in most of the nonpolar organic solvents. It is most commonly employed in organic reactions.Base employed in generating enolates for the preparation of lactone precursors, pyranones, and cyclohexanes.Used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines. Also used in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles.

Specifications

CAS 4039-32-1
Density 0.860 g/mL at 25°C
Molecular Formula C6H18LiNSi2
Linear Formula [(CH3)3Si]2NLi
MDL Number MFCD00008261
Quantity 250 g
Synonym Hexamethyldisilazane lithium salt
Molecular Weight (g/mol) 167.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.