Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Bis(Trimethylsilyl)Amide Solution

Catalog No. 766917100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-691-7100ML 800 mL
76-691-7800ML 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-691-7100ML Supplier Sigma Aldrich Supplier No. 766917100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Strong base for kinetically controlled deprotonations.

Specifications

CAS 4039-32-1
Density 0.893 g/mL at 25°C
Molecular Formula C6H18LiNSi2
Linear Formula [(CH3)3Si]2NLi
Quantity 800 mL
MDL Number MFCD00008261
Synonym Hexamethyldisilazane lithium salt
Molecular Weight (g/mol) 167.33 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.