Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Bis(Trimethylsilyl)Amide Solution

Catalog No. 577928800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-792-8800ML 800 mL
57-792-8100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 57-792-8800ML Supplier Sigma Aldrich Supplier No. 577928800ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lithium bis(trimethylsilyl)amide solution (LiHMDS) is generally used in organic synthesis as a non-nucleophilic strong Bronsted base.Lithium bis(trimethylsilyl)amide is generally used in organic synthesis as a non-nucleophilic strong Br++nsted base. It can be used for salt metathesis reaction for the synthesis of cesium bis(trimethylsilyl)amide (CsHMDS) and lithium fluoride by reacting with cesium fluoride.

Specifications

CAS 4039-32-1
Density 0.860 g/mL at 25°C
Molecular Formula C6H18LiNSi2
Linear Formula [(CH3)3Si]2NLi
MDL Number MFCD00008261
Quantity 800 mL
Synonym LiHMDS; Hexamethyldisilazane lithium salt
Molecular Weight (g/mol) 167.33 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.