Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lithium Borohydride Solution

Catalog No. 702714100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-271-4100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 70-271-4100ML Supplier Sigma Aldrich Supplier No. 702714100ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of gallium, indium, rhenium and zinc tris(mercaptoimidazolyl)hydroborato complexesMechano-chemical metathesis reactionsNoncatalytic hydrolysis for hydrogen generationGrowth of large gold monolayer protected-clustersAnion substitution reactionsDehydrogenation reactionsMaterial may form precipitate on standing, which does not affect its use.

Specifications

CAS 1218790-86-3
Melting Point 90°C to 94°C
Molecular Formula C9H13BN2O3
Linear Formula C9H13BN2O3
MDL Number MFCD11867876
Quantity 1 g
Molecular Weight (g/mol) 208.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.